Would you like to log in again?
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
Supplier: MilliporeSigma
1.10092.0021 1.10020.0001 1.10020.0001 1.10020.0002
EM1.10020.0001CS 12718.3 USD InStock
EM1.10020.0001 CA1.10092.0021 CA1.10020.0002 CA1.10020.0001
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
Environmental Test Kits
The nitrate test can be used for monitoring of drinking water, process water and wastewater, as well as aquarium water where limits can quickly be exceeded
MilliporeSigma # 1.10020.0001 replaces and is identical to MilliporeSigma # 10020-1
It can also be used for determine nitrate in food e.g. potatoes.
Accessories information: Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty