Would you like to log in again?
Peracetic Acid Test Method: Colorimetric With Test Strips 5 - 10 - 20 - 30 - 50 mg/L MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10084.0001 1.10084.0001
EM1.10084.0001CS 24948 USD InStock
CA1.10084.0001 EM1.10084.0001
Peracetic Acid Test Method: Colorimetric With Test Strips 5 - 10 - 20 - 30 - 50 mg/L MQuant™, MilliporeSigma
Environmental Test Kits
The test is suitable for the selective determination of peracetic acid traces in aqueous solutions and for check the absence of peracetic acid after rinsing processes, also in cases in which hydrogen peroxide is present.
MilliporeSigma # 1.10084.0001 replaces and is identical to MilliporeSigma # 10084-1
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty