Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®

Supplier: MilliporeSigma
Warning
Synonyms: Muriatic acidHydrochloric acid solution

1.10164.9950 1.10164.1000 1.10164.9025 1.10164.9950 1.10164.2500 1.10164.9208 1.10164.9025 1.10164.2500
EM1.10164.9208EA 3842 USD InStock
CA1.10164.9950 EM1.10164.9208 EM1.10164.1000 EM1.10164.2500 CA1.10164.9025 EM1.10164.9025 EM1.10164.9950 CA1.10164.2500
Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®
Hydrochloric acid
Manufactured from hydrochloric acid fuming 37% EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF and purified water acc. Ph. Eur., JP.

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Formula: HCl
MW: 36.46 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Concentration (HCl) 5.7 - 6.3 mol/l
Identity Passes Test
Color ≤10 Hazen
Appearance Passes Test
Free Bromine or Free Chlorine Passes Test
Free Chlorine (Cl) ≤0.0001
Sulfate (SO₄) ≤0.0005%
Sulfite (SO₃) ≤0.001%
Heavy Metals (as Pb) ≤0.0001%
As (Arsenic) ≤0.00015%
Residual Solvents (ICH Q3C) Excluded by Production Process
Residue on Ignition (as SO₄) ≤0.008%
Evaporation Residue ≤0.01
Endotoxins ≤0.25 I.U./ml
Filtered with 0.2 µm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty