Synonyms:
Muriatic acid
1.10164.9950
1.10164.1000
1.10164.9025
1.10164.9950
1.10164.2500
1.10164.9208
1.10164.9025
1.10164.2500
EM1.10164.9208EA
3842
USD
InStock
CA1.10164.9950
EM1.10164.9208
EM1.10164.1000
EM1.10164.2500
CA1.10164.9025
EM1.10164.9025
EM1.10164.9950
CA1.10164.2500
Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®
Hydrochloric acid
Manufactured from hydrochloric acid fuming 37% EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF and purified water acc. Ph. Eur., JP.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).