Would you like to log in again?
Peroxide Test Method: Colorimetric With Test Strips 100 - 200 - 400 - 600 - 800 - 1000 mg/L H₂O₂ MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10337.0001 1.10337.0001
EM1.10337.0001CS 32462 USD InStock
EM1.10337.0001 CA1.10337.0001
Peroxide Test Method: Colorimetric With Test Strips 100 - 200 - 400 - 600 - 800 - 1000 mg/L H₂O₂ MQuant™, MilliporeSigma
Environmental Test Kits
The peroxide test is suitable for checking the current concentration of diluted peroxide solutions to ensure the disinfectant effect.
MilliporeSigma # 1.10337.0001 replaces and is identical to MilliporeSigma # 10337-1
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty