Would you like to log in again?
Bacillus Subtilis (BGA) Spore Suspension, MilliporeSigma
Supplier: MilliporeSigma
1.10649.0001
EM1.10649.0001EA 176.97 USD InStock
EM1.10649.0001
Bacillus Subtilis (BGA) Spore Suspension, MilliporeSigma
Media for Microbiology Additives and Supplements for Microbiology
For inhibitor tests.
  • 8E+06 - 5E+07 CFU/mL viable spores in Bacillus subtilis DSM 618
  • Passes performance test
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty