Would you like to log in again?
MColortest™ Oxygen Test, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.11152.0001 1.11152.0001
EM1.11152.0001EA 343 USD InStock
EM1.11152.0001 CA1.11152.0001
MColortest™ Oxygen Test, MilliporeSigma
Environmental Test Kits
The sustainability of biological processes in natural waters is essentially depended on a sufficient oxygen content
When the reagents of the MColortestTM test are used up, the kit can be refilled by ordering the appropriate refill pack. They are available for a range of MColortestTM test kits, making it a particularly economical system.
The oxygen present in water is also consumed by chemical processes and by microorganisms.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty