Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BACTIDENT® Indole Reagent (Kovac’s Reagent), MilliporeSigma
Supplier: MilliporeSigma
1.11350.0001 1.11350.0001 1.13301.0001 1.13301.0001
EM1.11350.0001EA 53.45 USD InStock
CA1.11350.0001 CA1.13301.0001 EM1.11350.0001 EM1.13301.0001
BACTIDENT® Indole Reagent (Kovac’s Reagent), MilliporeSigma
Microbiology Test Systems Endotoxin Tests
EMD Product # 1.13301.0001 replaces and is identical to EMD Product # 1.13301.0002.
These test kits are most commonly used to aid in the identification of bacterial colonies. Use of these products allows for the streamlining of testing protocols, ensuring the safety and reliability of finished products, and shortening of overall production cycles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty