Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming 37%, EMSURE® for analysis (max. 0.001 ppm Hg), Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Muriatic acid

M-Clarity™ quality level = MQ300

1.13386.2500 1.13386.2500
EM1.13386.2500CS 1166 USD InStock
CA1.13386.2500 EM1.13386.2500
Hydrochloric acid fuming 37%, EMSURE® for analysis (max. 0.001 ppm Hg), Supelco®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Al (Aluminium) ≤ 0.05 ppm
As (Arsenic) ≤ 0.01 ppm
Assay (alkalimetric) ≥ 37.0 %
Ba (Barium) ≤ 0.02 ppm
Be (Beryllium) ≤ 0.02 ppm
Bi (Bismuth) ≤ 0.1 ppm
Bromide (Br) ≤ 50 ppm
Ca (Calcium) ≤ 0.5 ppm
Cd (Cadmium) ≤ 0.01 ppm
Co (Cobalt) ≤ 0.01 ppm
Color ≤ 10 Hazen
Cr (Chromium) ≤ 0.02 ppm
Cu (Copper) ≤ 0.02 ppm
Fe (Iron) ≤ 0.2 ppm
Free chlorine (Cl) ≤ 0.5 ppm
Ge (Germanium) ≤ 0.05 ppm
Hg (Mercury) ≤ 0.001 ppm
K (Potassium) ≤ 0.1 ppm
Li (Lithium) ≤ 0.01 ppm
Mg (Magnesium) ≤ 0.1 ppm
Mn (Manganese) ≤ 0.01 ppm
Mo (Molybdenum) ≤ 0.02 ppm
Na (Sodium) ≤ 0.5 ppm
NH₄ (Ammonium) ≤ 1 ppm
Ni (Nickel) ≤ 0.02 ppm
Pb (Lead) ≤ 0.02 ppm
Phosphate (PO₄) ≤ 0.5 ppm
Residue on ignition (as SO₄) ≤ 5 ppm
Sb (Antimony) ≤ 0.002 ppm
Sr (Strontium) ≤ 0.01 ppm
Sulfate (SO₄) ≤ 1 ppm
Sulfite (SO₃) ≤ 1 ppm
Ti (Titanium) ≤ 0.1 ppm
Tl (Thallium) ≤ 0.05 ppm
V (Vanadium) ≤ 0.01 ppm
Zn (Zinc) ≤ 0.05 ppm
Zr (Zirconium) ≤ 0.1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty