Would you like to log in again?
MColortest™ Chromate Test, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.14441.0001 1.14402.0001 1.14441.0001 1.14402.0001
EM1.14402.0001EA 265 USD InStock
CA1.14402.0001 EM1.14402.0001 CA1.14441.0001 EM1.14441.0001
MColortest™ Chromate Test, MilliporeSigma
Environmental Test Kits
The test measures chromium(VI) present in the sample as chromate or dichromate ions in wastewater and percolating water, groundwater and surface water, industrial water and drinking water.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty