Would you like to log in again?
Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.010 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/L Cl₂ MColortest™
Supplier: MilliporeSigma
MColortest™
1.14434.0001 1.14434.0001
EM1.14434.0001CS 3697 USD InStock
EM1.14434.0001 CA1.14434.0001
Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.010 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/L Cl₂ MColortest™
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty