Would you like to log in again?
Spectroquant® Nitrogen (total) Cell Test Method: photometric 0.5 - 15.0 mg/L N
Supplier: MilliporeSigma
Spectroquant®
1.14537.0001 1.14537.0001
EM1.14537.0001CS 1496 USD InStock
CA1.14537.0001 EM1.14537.0001
Spectroquant® Nitrogen (total) Cell Test Method: photometric 0.5 - 15.0 mg/L N
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty