Would you like to log in again?
Silicate (Silicic Acid) Test, Spectroquant®, Supelco, MilliporeSigma
Supplier: MilliporeSigma
Spectroquant®
1.14794.0007
EM1.14794.0007EA 471 USD InStock
EM1.14794.0007
Silicate (Silicic Acid) Test, Spectroquant®, Supelco, MilliporeSigma
Environmental Test Kits
Method: photometric 0.011 - 10.70 mg/l SiO₂ 0.005 - 5.00 mg/l Si Spectroquant® with 10 mm cell
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty