Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin B, Certistain® for microscopy (C.I. 45400), Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 2,7-Dinitro-4,5-dibromofluoresceinEosin bluishEosin B disodium saltAcid Red 912-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt

1.15934.0025 1.15934.0025
EM1.15934.0025CS 11048.75 USD InStock
CA1.15934.0025 EM1.15934.0025
Eosin B, Certistain® for microscopy (C.I. 45400), Sigma-Aldrich®
Eosin B


Formula: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
MDL Number: MFCD00005041
CAS Number: 548-24-3
Order Now
Specification Test Results
Solubility 300 g/l (25 °C)
Molar Mass 624.06 g/mol
Bulk Density 680 kg/m3
pH 6.2 (10 g/l, H2O, 25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty