Would you like to log in again?
Reflectoquant® Lactic Acid Test, MilliporeSigma
Supplier: MilliporeSigma
1.16127.0001 1.16127.0001
EM1.16127.0001CS 46971 USD InStock
EM1.16127.0001 CA1.16127.0001
Reflectoquant® Lactic Acid Test, MilliporeSigma
Environmental Test Kits
The test measures total lactic acid (sum of D- and L-lactic acid in beverages, e.g. wine, beer and fruit/vegetable juice after dilution. Furthermore the test can be used for the determination of lactic acid in food after pretreatment. Enzymatic reaction.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty