Would you like to log in again?
Blank Test Strips Without Reagents Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
Reflectoquant®
1.16730.0001 1.16730.0001
EM1.16730.0001CS 12422 USD InStock
CA1.16730.0001 EM1.16730.0001
Blank Test Strips Without Reagents Reflectoquant®, MilliporeSigma
Environmental Test Kits
When analyzing coloured samples using the Reflectoquant® System the intrinsic colouration can, in given circumstances, produce more or less pronounced false-high results.
Therefore the blank strips can be used to correct these measurement values without having to take time-consuming sample-pretreatment measures.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty