Would you like to log in again?
Total Hardness Test Method: Reflectometric With Test Strips 0.1 - 30.0 °d Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
Reflectoquant®
1.16997.0001 1.16997.0001
EM1.16997.0001CS 36266 USD InStock
EM1.16997.0001 CA1.16997.0001
Total Hardness Test Method: Reflectometric With Test Strips 0.1 - 30.0 °d Reflectoquant®, MilliporeSigma
Environmental Test Kits
Test strips for the determination of water hardness with reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Test kits with reagents for reliable documented quantitative results
  • Barcode calibration
  • Low analysis cost
  • Test strips with minute amount of chemicals for easy disposal
The test measures the content of salts of the alkaline earth metals calcium, magnesium, strontium and barium in groundwater and surface water, drinking water, mineral water and boiler water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty