Would you like to log in again?
Peracetic Acid Test Method: Colorimetric With Test Strips 500 - 1000 - 1500 - 2000 mg/L MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.17922.0001 1.17922.0001
EM1.17922.0001CS 25976 USD InStock
CA1.17922.0001 EM1.17922.0001
Peracetic Acid Test Method: Colorimetric With Test Strips 500 - 1000 - 1500 - 2000 mg/L MQuant™, MilliporeSigma
Environmental Test Kits
The peracetic acid tests for higher concentrations of peracetic acid are eminently suitable for checking that the prescribed end concentration of peracetic acid-based detergents are being maintained.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty