Would you like to log in again?
Nitrate Test RQeasy® Method: Reflectometric 5 - 250 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Supplier: MilliporeSigma
RQeasy®
1.17961.0001 1.17961.0001
EM1.17961.0001CS 20044.45 USD InStock
CA1.17961.0001 EM1.17961.0001
Nitrate Test RQeasy® Method: Reflectometric 5 - 250 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Environmental Test Kits
The rapid determination of the nitrate content is of particular interest with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g baby foods).
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty