Would you like to log in again?
Turbiquant® 1100 Portable Turbidity Meters, Merck Millipore
Supplier: MilliporeSigma
1.18324.0001 1.18324.0001
EM1.18324.0001EA 2528 USD InStock
CA1.18324.0001 EM1.18324.0001
Turbiquant® 1100 Portable Turbidity Meters, Merck Millipore
Turbidity Meters
The degree of turbidity is a measure of water quality - A liquid is cloudy if it contains undissolved suspended matter such as particles or bacteria.
  • Battery power for over 5000 measurements
  • Improved precision, ready to use and non toxic standards
  • Portable battery powered for on the spot analysis
These particles scatter incident light in many directions. The light scattered at 90 degrees is measured. Nephelometry is the measurement of turbidity. The clearer a liquid, the lower the turbidity. The units of turbidity are nephelometric turbidity units (NTU). Drinking water has max. of 5 NTU, sewage more than 2000 NTU.
Certifications: ISO 7027 an IR LED (infrared) light source with a wavelength of 860 nm is prescribed (IR models). USEPA method 180.1 and also APHA-AWWA-WPCF require the use of a tungsten lamp emitting white light (T models). AMCO-AEPA-1® microspheres are primary standards conform with USEPA method 180.1. In compliance with EN ISO 7027, these standards count as alternative secondary standards to fresh formazine.
Delivery information: Each Turbiquant requires the appropriate set of calibration standards to be purchased separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty