Would you like to log in again?
Visual Color Kits, Sulfate, MColortest™, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.18389.0001 1.18389.0001
EM1.18389.0001CS 2841 USD InStock
CA1.18389.0001 EM1.18389.0001
Visual Color Kits, Sulfate, MColortest™, MilliporeSigma
Environmental Test Kits
Test kits for quick, visual colour comparison, semi-quantitative analysis in situ, without the expense of transporting samples to the laboratory. Simple step by step illustrated instructions mean the kits can be used without special chemical training. The various systems differ in their format, application and sensitivity.
  • Easy to use, results in approximately 1 minute
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, color comparison chart or comparator wheel and extensive instructions for use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty