Would you like to log in again?
MColortest™ Iron Test, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.14438.0001 1.18458.0002 1.14438.0001 1.14759.0001 1.18458.0002 1.14759.0001
EM1.14759.0001EA 452 USD InStock
EM1.14759.0001 EM1.18458.0002 CA1.14438.0001 EM1.14438.0001 CA1.18458.0002 CA1.14759.0001
MColortest™ Iron Test, MilliporeSigma
Environmental Test Kits
The test measures bivalent and trivalent iron in its dissolved form as well as fresh colloidal iron(III) hydroxide in groundwater and surface water, drinking water, industrial water and wastewater.
When the reagents of the MColortest™ test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortest™ test kits, making it a particularly economical system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty