Would you like to log in again?
Fluoride Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.18771.0001 1.18771.0001
EM1.18771.0001EA 390 USD InStock
CA1.18771.0001 EM1.18771.0001
Fluoride Test, MQuant®, MilliporeSigma
Environmental Test Kits
Quick tests, visual colour comparison
  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty