Synonyms:
Caustic sodaLyeSodium hydrateSoda lye
1.37020.5000
1.37020.1000
1.37020.9012
EM1.37020.9012EA
1246
USD
InStock
EM1.37020.9012
EM1.37020.5000
EM1.37020.1000
Sodium hydroxide ≥98.0% (by acidimetry), pellets, EMPROVE® EXPERT Ph. Eur., BP, JP, NF for biopharmaceutical production, SAFC®
Sodium hydroxide
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Certifications: conforms to Ph. Eur., BP, JP, NF