Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥98.0% (by acidimetry), pellets, EMPROVE® EXPERT Ph. Eur., BP, JP, NF for biopharmaceutical production, SAFC®

Supplier: MilliporeSigma
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

1.37020.5000 1.37020.1000 1.37020.9012
EM1.37020.9012EA 1246 USD InStock
EM1.37020.9012 EM1.37020.5000 EM1.37020.1000
Sodium hydroxide ≥98.0% (by acidimetry), pellets, EMPROVE® EXPERT Ph. Eur., BP, JP, NF for biopharmaceutical production, SAFC®
Sodium hydroxide
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Certifications: conforms to Ph. Eur., BP, JP, NF


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Order Now
Specification Test Results
Assay (acidimetric,NaOH) ≥ 98.0 %
Assay (total alkalinity calc. as NaOH) 98.0 - 100.5 %
Assay (Na) 54.0 - 59.8 %
Identity passes test
Appearance of solution passes test
Insoluble substances and organic matter passes test
Carbonate (as Na₂CO₃) ≤ 0.5 %
Chloride (Cl) ≤ 0.015 %
Phosphat (PO4) ≤ 0.001 %
Silicate (SiO₂) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.010 %
Total nitrogen (N) ≤ 0.0005 %
Heavy metals (as Ag) ≤ 0.002 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.001 %
As (Arsenic) ≤ 0.0003 %
Ca (Calcium) ≤ 0.005 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.0010 %
Hg (Mercury) ≤ 0.00001 %
K (Potassium) ≤ 0.05 %
Mg (Magnesium) ≤ 0.002 %
Ni (Nickel) ≤ 0.001 %
Pb (Lead) ≤ 0.00005 %
Zn (Zinc) ≤ 0.0025 %
Residual solvents (Ph Eur/USP/ICH) excluded by manufacturing process
Endotoxins ≤ 2.5 I.U./g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty