Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®

Supplier: MilliporeSigma
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

Filtered through a 0.2 µm filter.

1.37023.5000 1.37023.5000 1.37023.2500 1.37023.9200 1.37023.9025 1.37023.9950 1.37023.9025 1.37023.9200
EM1.37023.5000CS 2020 USD InStock
CA1.37023.5000 EM1.37023.5000 EM1.37023.9200 EM1.37023.9025 EM1.37023.2500 CA1.37023.9200 CA1.37023.9025 CA1.37023.9950
Sodium hydroxide solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide
This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph Eur.

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Formula: NaOH
MW: 40 g/mol
Melting Pt: 9 °C
Density: 1.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,
Order Now
Specification Test Results
Assay (acidimetric,NaOH) 32.0 - 33.0 %
Carbonate (as Na₂CO₃) ≤ 1.0 %
Chloride (Cl) ≤ 5 ppm
Sulfate (SO₄) ≤ 30 ppm
Heavy metals (as Pb) ≤ 5.0 ppm
Al (Aluminium) ≤ 10 ppm
Fe (Iron) ≤ 3 ppm
Ni (Nickel) * ≤ 2.00 ppm
Residual solvents (ICH Q3C) ...excluded by production process
Endotoxins ≤ 2.0 I.U./ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty