Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1 mol/L, EMPROVE® EXPERT

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

1.37031.2500 1.37031.1002 1.37031.2500 1.37031.9025 1.37031.9200 1.37031.9200 1.37031.9958 1.37031.9025 1.37031.9208
EM1.37031.9025EA 956 USD InStock
CA1.37031.2500 EM1.37031.9025 EM1.37031.9200 EM1.37031.9958 EM1.37031.1002 CA1.37031.9025 EM1.37031.2500 CA1.37031.9200 EM1.37031.9208
Sodium hydroxide 1 mol/L, EMPROVE® EXPERT
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Density: 1.04 g/cm³ (20 °C)
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Assay (Acidimetric, NaOH) 0.99 - 1.01 mol/l
Carbonate (as Na₂CO₃) ≤1.0%
Chloride (Cl) ≤5 ppm
Sulfate (SO₄) ≤30 ppm
Heavy Metals (as Pb) ≤5.0 ppm
Al (Aluminium) ≤10 ppm
Fe (Iron) ≤3 ppm
Residual Solvents (ICH Q3C) Excluded by Production Process
Endotoxins ≤2.0 I.U./ml
Total Aerobic Microbial Count (TAMC) ≤100 CFU/g
Total Combined Yeasts/Moulds Count (TYMC) ≤10 CFU/g
Filtered with 0.2 µm
This product is manufactured from sodium hydroxide pellets Ph. Eur., BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph. Eur.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty