Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
Filtered through a 0.2 µm filter.
1.37058.0001
1.37058.9025
1.37058.9950
EM1.37058.9950EA
11677
USD
InStock
CA1.37058.0001
EM1.37058.9950
EM1.37058.9025
Sodium hydroxide 0.1 mol/L in aqueous solution, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide
Suitable for biopharmaceutical production.
This product has been manufactured from Sodium hydroxide pellets Ph. Eur., BP, JP, NF and waterfor injection (wfi) ace. USP which conforms with highly purified water (hpw) ace. Ph Eur.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elementallmpurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).