Would you like to log in again?
COLUMN 100 RP-18 LICHROSORB HIBAR SS 5UM
Supplier: MilliporeSigma
LiChrospher* 100 RP-18 endcapped Hibar* column 5um^ LiChrospher* 100 RP-18 endcapped Hibar* column, Material: stainless steel, Particle size: 5 um, Sorbent characteristics: Particles of silica with octadecyl derivative endcapped, Pore size: 100 nm, Length: 125 mm, I.D: 4.6 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN 100 RP-18 LICHROSORB HIBAR SS 5UM EM1.51906.0001 1.51906.0001 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty