Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol ≥96% (v/v), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ethyl alcohol

M-Clarity™ quality level = MQ300

1.59010.2500 1.59010.0500
EM1.59010.0500CS 1694 USD InStock
EM1.59010.0500 EM1.59010.2500
Ethanol ≥96% (v/v), EMSURE® for analysis, Supelco®
Ethanol
Ethanol 96% EMSURE® Reag. Ph Eur.


Formula: C₂H₅OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,
Order Now
Specification Test Results
Grade Reag. Ph Eur
Hill Formula C₂H₆O
Chemical formula C₂H₅OH
Molar Mass 46.07 g/mol
Boiling point 78 °C (1013 hPa)
Density 0.805 - 0.812 g/cm³ (20 °C)
Explosion limit 3.5 - 15 %(V)
Flash point 17 °C
Ignition temperature 425 °C
Melting Point -117 °C
pH value 7.0 (10 g/l, H₂O, 20 °C)
Vapor pressure 59 hPa (20 °C)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 6200 mg/kg
Storage class 3 Flammable liquids
WGK WGK 1 slightly water endangering
R Phrase R 11
S Phrase S 7-16

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty