Would you like to log in again?
Spectroquant® Move 100 Colorimeter, MilliporeSigma
Supplier: Merck
Spectroquant®
1.73632.0001 1.73632.0001
EM1.73632.0001EA 2044 USD InStock
CA1.73632.0001 EM1.73632.0001
Spectroquant® Move 100 Colorimeter, MilliporeSigma
Colorimeters Multiparameter Colorimeters
The Spectroquant® Move Cl2/O3/ClO2/CyA/pH mobile colorimeter is suitable for the detection of five essential parameters of water analysis: chlorine, ozone, chlorine dioxide, cyanuric acid and pH
  • More than 100 high quality Spectroquant® tests pre-programmed - covers all the important methods for routine analysis
  • IP 68, dust tight and waterproof
  • Accurate instrument: ±1 nm, filters with Δλ =5 (typical)
  • Acid and solvent resistant tactile film keypad with acoustic feedback
  • Auto off, 20 minutes after the last function
  • 8 language options
This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.
Ordering information: Standard contents of the Spectroquant Colorimeter Move 100 include the colorimeter in a plastic carrying case, 4 batteries (type AA/LR6), adapter for 16mm dia. round cells, 3 round cells with 16mm dia. cap, 3 round cells with 24mm dia. cap, screwdriver, operating instructions manual, and certificate of compliance.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty