Would you like to log in again?
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
AQUASTAR®
1.88011.1045 1.88009.1000 1.88015.2500 1.88015.1000 1.88016.1000 1.09255.0500 AX1697C-30 1.09257.0500 1.88009.2500 1.88001.1045 1.88007.1002 1.88015.2500 1.88010.1045 AX1697C-1 1.09257.0500 1.88005.2500 AX1697E-1 1.88005.1000 1.88010.1045 1.88005.2500 AX1697E-1 1.88005.1003 1.09255.0500 AX1697C-30 1.88001.1045 1.88011.1045 AX1697A-1 AX1697C-1 1.88020.1000 1.88009.2500 1.88002.1045 AX1697E-1 1.88016.1000 1.88008.1000 AX1697A-1 1.88015.1000 AX1698E-6 1.88009.1000 AX1697F-1 1.88005.0500 1.88005.0500 1.88006.1000
EM-1.88016.100EA 149 USD InStock
CA1.88011.1045 EM-1.88016.100 EM1.09257.0500 EM1.88005.1003 EM1.88002.1045 EM1.88010.1045 CA1.88006.1000 EM1.88015.2500 EM1.09255.0500 EM1.88007.1002 CA1.88016.1000 CA1.88015.2500 CA1.88008.1000 EM-AX1697C-30 CA1.88009.1000 EM-AX1697A-1 EM-AX1697C-1 CA1.88010.1045 EM-AX1697E-1 EM1.88011.1045 EM1.88005.0500 EM1.88005.2500 EM188009.1000 CA1.88009.2500 CA1.09257.0500 CA1.88001.1045 EM188015.1000 CAAX1697E-1 CAAX1697F-1 CAAX1697C-30 EM1.88001.1045 CA1.88020.1000 CA1.88005.2500 CAAX1697A-1 CAAX1697C-1 CA-AX1697E-1 EM188009.2500 CA1.88015.1000 EM1.88005.1000 CA1.09255.0500 CA1.88005.0500 EM-AX1698E-6
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Karl Fischer Titrants and Reagents
The Aquastar® range is designed to provide reagents for the determination of water by Karl Fischer titration of a wide range of substrates and by a variety of methodologies. These include reagents for one- or two-component systems; combined coulometric reagents; solvents for oils and fats; reagents and solvents for determination of water in ketones and aldehydes. Calibration standards are also available.
  • CombiTitrants and CombiSolvents are non toxic
  • Very accurate and reproducible results
  • Rapid and reliable
Merck have introduced into the range of Aquastar® Titrants, some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty