Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 10% (w/v), EMPROVE® EXPERT Ph. Eur., JP, NF, SAFC®

Supplier: MilliporeSigma
Warning
Synonyms: Muriatic acidHydrochloric acid solution

4.80592.9958 4.80592.9208 4.80592.9950 4.80592.9026 4.80592.2500 4.80592.9200
EM4.80592.9208EA 4846 USD InStock
EM4.80592.9208 CA4.80592.9950 CA4.80592.9026 CA4.80592.9200 EM4.80592.9958 EM4.80592.2500
Hydrochloric acid 10% (w/v), EMPROVE® EXPERT Ph. Eur., JP, NF, SAFC®
Hydrochloric acid
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

  • Filtered with 0.2 µm

Certifications: Corresponds to Ph. Eur., JP, NF.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 102 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Assay (alkalimetric) 9.5 - 10.0 %
Assay (alkalimetric) 10.0 - 10.5 % (w/v)
Identity passes test
Color ≤ 10 Hazen
Appearance passes test
Bromide or iodide passes test
Free bromine or chlorine passes test
Free chlorine (Cl) ≤ 0.0001
Sulfate (SO₄) ≤ 0.0005 %
Sulphite (SO3) ≤ 0.001 %
Heavy metals (as Pb) ≤ 0.0001 %
As (Arsenic) ≤ 0.000015 %
Hg (Mercury) ≤ 0.0000010 %
Residual solvents (ICH Q3C) ...excluded by production process
Evaporation residue ≤ 0.01
Residue on ignition (as SO₄) ≤ 0.008 %
Endotoxins ≤ 2.5 I.U./ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty