Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

M-Clarity™ quality level = MQ200

4.90061.0100 4.90061.0025 4.90061.0025 4.90061.0100
EM4.90061.0025CS 27271 USD InStock
CA4.90061.0100 CA4.90061.0025 EM4.90061.0025 EM4.90061.0100
1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate for synthesis, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate


Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
MDL Number: MFCD00210008
CAS Number: 145022-44-2
Order Now
Specification Test Results
Melting Point -12 °C
Molar Mass 260.24 g/mol
Density 1.386 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty