Would you like to log in again?
S-protein agarose, MilliporeSigma
Supplier: MilliporeSigma
69704-3 69704-3 69704-4 69704-4
EM69704-3EA 214.16 USD InStock
CA80031-014 EM69704-3 EM69704-4 CA80031-016
S-protein agarose, MilliporeSigma
Protein Purification Protein Purification Resins
S-protein agarose specifically retains STag™ fusion proteins. The capacity of S-protein agarose varies and is based on the size and folding characteristics of a given target protein.
Binding can be performed in a column or batch mode. Elution is possible using slightly denaturing conditions or using a specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty