Would you like to log in again?
BugBuster® 10X Protein Extraction Reagent, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70921-4 70921-5 70921-4 70921-5
EM70921-4EA 668.69 USD InStock
EM70921-4 CA80509-636 EM70921-5 CA80508-846
BugBuster® 10X Protein Extraction Reagent, MilliporeSigma
Protein Extraction Protein Extraction Reagents
BugBuster 10X is a concentrated formulation of the proprietary detergents employed in BugBuster without the addition of salts or buffer components.
  • Bioprocessing benefits
  • Freedom to control pH
  • Maximum extraction and activitiy of your target protein
  • Allows user-defined dilution
This has all of the bioprocessing benefits of standard BugBuster in addition to the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein. Concentrated BugBuster provides a flexible alternative to the ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty