Would you like to log in again?
Lysonase™ Bioprocessing Reagent, EMD Millipore
Supplier: MilliporeSigma
71230-3 71230-5 71230-4 71230-4
EM71230-4EA 830.9 USD InStock
CA82601-956 CA82601-952 EM71230-4 CA82601-954
Lysonase™ Bioprocessing Reagent, EMD Millipore
Protein Extraction Protein Extraction Reagents
Lysonase™ bioprocessing reagent is an optimized, ready-to-use blend of rLysozyme™ solution and benzonase nuclease.
  • Increased protein extraction efficiency
  • Facilitate downstream processing of protein extracts
rLysozyme solution contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme. Benzonase nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear), reducing extract viscosity and increasing protein yield.
For efficient protein extraction with BugBuster® protein extraction reagent, use 10 µl Lysonase per 1 g cell paste. For efficient protein extraction with PopCulture® reagent, add 2 µl Lysonase per 1 ml culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty