Would you like to log in again?
D-Desthiobiotin, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Novagen®
71610-3 71610-3
EM71610-3EA 629.48 USD InStock
CA80017-072 EM71610-3
D-Desthiobiotin, MilliporeSigma
Protein Purification Protein Purification Resins
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty