Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminocaproic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: ε-Amino-n-caproic acidH-Eahx-OHAminocaproic Acid6-Aminocaproic Acid6-Aminohexanoic acid

M-Clarity™ quality level = MQ200

8.00145.0250 8.00145.5000 8.00145.1000 8.00145.0250 8.00145.0005 8.00145.5000 8.00145.1000 8.00145.9010 8.00145.0005 8.00145.9010
EM8.00145.0005CS 32018 USD InStock
CA8.00145.0250 CA8.00145.1000 EM8.00145.0005 CA8.00145.5000 EM8.00145.1000 CA8.00145.9010 EM8.00145.5000 EM8.00145.0250 CA8.00145.0005 EM8.00145.9010
6-Aminocaproic acid for synthesis, Sigma-Aldrich®
6-Aminocaproic acid


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 127 °C
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431
Order Now
Specification Test Results
Solubility 800 g/l (20 °C)
Molar Mass 131.17 g/mol
Flash Point 207 - 209 °C
pH 7.0 - 7.5 (50 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty