Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acrylic acid stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Prop-2-enoic acid

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00181.0100 8.00181.0100 8.00181.1000 8.00181.0500 8.00181.1000
EM8.00181.1000CS 530 USD InStock
EM8.00181.1000 EM8.00181.0100 EM8.00181.0500 CA8.00181.1000 CA8.00181.0100
Acrylic acid stabilized for synthesis, Sigma-Aldrich®
Acrylic acid


Formula: C₃H₄O₂
MW: 72.06 g/mol
MDL Number: MFCD00004367
CAS Number: 79-10-7
UN: 2218
ADR: 8 (3),II
Order Now
Specification Test Results
Solubility 1000 g/l (25 °C)
Melting Point 13 °C
Molar Mass 72.06 g/mol
Boiling Point 141 °C (1013 hPa)
Vapor Pressure 3.8 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.4224 (20 °C, 589 nm)
Explosion Limit 3.9 - 19.8 %(V)
Density 1.05 g/cm3 (20 °C)
pH 2.1 (72.06 g/l, H2O, 20 °C)
Ignition Point 390 °C DIN 51794
Synonyms 2-Propenoic acid, Acroleic acid
Hill Formula C3H4O2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty