Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caproic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Hexanoic acid

M-Clarity™ quality level = MQ200

8.00198.0005 8.00198.0250 8.00198.0005 8.00198.1000 8.00198.1000 8.00198.0250
EM8.00198.0250CS 466 USD InStock
EM8.00198.0250 EM8.00198.1000 CA8.00198.0250 EM8.00198.0005 CA8.00198.1000 CA8.00198.0005
Caproic acid for synthesis, Sigma-Aldrich®
Caproic acid


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –4.8 °C
Density: 0.9276 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00004421
CAS Number: 142-62-1
EINECS: 205-550-7
UN: 2829
ADR: 8,III
Merck Index: 13,01765
Order Now
Specification Test Results
Solubility 9.7 g/l (20 °C)
Melting Point -3 °C
Molar Mass 116.16 g/mol
Boiling Point 206 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4162 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
pH 4 (1 g/l, H2O, 20 °C)
Ignition Point 380 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty