Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl octanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Caprylic acid ethyl esterOctanoic acid ethyl esterEthyl caprylate

M-Clarity™ quality level = MQ200

8.00202.0005 8.00202.0250 8.00202.0005 8.00202.9010 8.00202.0250
EM8.00202.0250CS 641 USD InStock
EM8.00202.0250 CA8.00202.0005 EM8.00202.9010 EM8.00202.0005 CA8.00202.0250
Ethyl octanoate for synthesis, Sigma-Aldrich®
Ethyl octanoate


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Melting Pt: –47 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00009552
CAS Number: 106-32-1
ADR: 9,III
Order Now
Specification Test Results
Solubility 0.070 g/l (25 °C)
Melting Point -47 °C
Molar Mass 172.27 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.4166 (20 °C, 589 nm)
Explosion Limit 0.7 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 325 °C
Synonyms Ethyl caprylate, Octanoic acid ethyl ester
Hill Formula C10H20O2
Chemical Formula CH3(CH2)6COOC2H5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty