Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl cyanide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.00258.0025 8.00258.0025
EM8.00258.0025CS 9759 USD InStock
CA8.00258.0025 EM8.00258.0025
Allyl cyanide for synthesis, Sigma-Aldrich®
Allyl cyanide


Formula: C₄H₅N
MDL Number: MFCD00001962
CAS Number: 109-75-1
EINECS: 203-701-1
UN: 1992
ADR: 3 (6.1),III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point -87 °C
Molar Mass 67.09 g/mol
Boiling Point 116 - 121 °C
Flash Point 23 °C
Refractive Index 1.405 (20 °C, 589 nm)
Density 0.83 g/cm3 (25 °C)
Ignition Point 455 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty