Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Allyl-2,3-epoxypropyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Allyl glycidyl ether

M-Clarity™ quality level = MQ200

8.00259.2500 8.00259.2500 8.00259.0250 8.00259.0250
EM8.00259.0250CS 1240 USD InStock
EM8.00259.0250 CA8.00259.2500 EM8.00259.2500 CA8.00259.0250
(±)-Allyl-2,3-epoxypropyl ether for synthesis, Sigma-Aldrich®
(±)-Allyl 2,3-epoxypropyl ether


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00005143
CAS Number: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III
Order Now
Specification Test Results
Solubility 50 g/l (20 °C)
Melting Point -100 °C
Molar Mass 114.15 g/mol
Boiling Point 154 °C (1013 hPa)
Vapor Pressure 2.6 hPa (20 °C)
Flash Point 45 °C
Refractive Index 1.4332 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
Ignition Point 264 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty