Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Hydroxyaniline

M-Clarity™ quality level = MQ200

8.00419.9010 8.00419.0250 8.00419.0005 8.00419.0005 8.00419.0250 8.00419.1000 8.00419.1000
EM8.00419.9010EA 664 USD InStock
EM8.00419.9010 CA8.00419.0250 EM8.00419.0005 CA8.00419.1000 CA8.00419.0005 EM8.00419.0250 EM8.00419.1000
2-Aminophenol for synthesis, Sigma-Aldrich®
2-Aminophenol


Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00007690
CAS Number: 95-55-6
UN: 2512
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 17 g/l (20 °C)
Melting Point 172 - 174 °C (sublimed)
Molar Mass 109.12 g/mol
Bulk Density 600 kg/m3
Boiling Point 150 °C (15 hPa)
Vapor Pressure 14 hPa (153 °C)
Flash Point 175 °C
Density 1.328 g/cm3 (20 °C)
pH 6.5 - 7.5 (10 g/l, H2O, 20 °C)
Ignition Point 190 °C
Synonyms 2-Hydroxy aniline, 2-Amino-1-hydroxy benzene
Hill Formula C6H7NO
Chemical Formula 2-(NH2)C6H4OH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty