Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyl methacrylate stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Methylacrylic acid, butyl esterMethacrylic acid butyl estern-Butyl Methacrylate, MonomerButyl 2-methacrylate

M-Clarity™ quality level = MQ200

8.00585.0100 8.00585.0500 8.00585.0100 8.00585.0500
EM8.00585.0100CS 698 USD InStock
EM8.00585.0100 EM8.00585.0500 CA8.00585.0100 CA8.00585.0500
Butyl methacrylate stabilized for synthesis, Sigma-Aldrich®
Butyl methacrylate


Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –75 °C
Density: 0.8951 g/cm³ (20 °C)
MDL Number: MFCD00009444
CAS Number: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3,III
Order Now
Specification Test Results
Solubility 0.36 g/l (25 °C)
Melting Point -75 °C
Molar Mass 142.20 g/mol
Boiling Point 163 °C (1013 hPa)
Vapor Pressure 2.7 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.424 (20 °C, 589 nm)
Explosion Limit 2.00 - 8.00 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 290 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty