Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salicylaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Hydroxybenzaldehyde

M-Clarity™ quality level = MQ200

8.00640.9025 8.00640.0250 8.00640.9025 8.00640.1000 8.00640.0250 8.00640.0005 8.00640.1000 8.00640.0005
EM8.00640.1000EA 124 USD InStock
EM8.00640.1000 CA8.00640.0250 CA8.00640.1000 EM8.00640.0005 CA8.00640.9025 EM8.00640.9025 CA8.00640.0005 EM8.00640.0250
Salicylaldehyde for synthesis, Sigma-Aldrich®
Salicylaldehyde


Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003317
CAS Number: 90-02-8
EINECS: 201-961-0
UN: 3082
ADR: 9,III
Merck Index: 12,08478
Order Now
Specification Test Results
Solubility 5 g/l (20 °C)
Melting Point 224 - 255 °C
Molar Mass 167.12 g/mol
Bulk Density 700 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 188 °C
pH 2.0 (5 g/l, H2O, 20 °C)
Ignition Point 620 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty