Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrobenzoic acid, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.00646.9050 8.00646.9050
EM8.00646.9050EA 2926 USD InStock
CA8.00646.9050 EM8.00646.9050
4-Nitrobenzoic acid, Sigma-Aldrich®
4-Nitrobenzoic acid


Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 121 °C
Density: 1.65 g/cm³ (20 °C)
MDL Number: MFCD00007352
CAS Number: 62-23-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty