Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terephthalic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.00762.0100 8.00762.1000 8.00762.0100 8.00762.1000
EM8.00762.1000CS 509 USD InStock
CA8.00762.0100 EM8.00762.1000 EM8.00762.0100 CA8.00762.1000
Terephthalic acid for synthesis, Sigma-Aldrich®
Terephthalic acid


Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 392 °C (1013 hPa)
Density: 1.472 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Merck Index: 12,09306
Order Now
Specification Test Results
Assay (acidimetric) ≥ 98.0 %
Identity (IR) passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty