Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acrylamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Prop-2-enamide

M-Clarity™ quality level = MQ200

8.00830.5000 8.00830.0100 8.00830.0100 8.00830.5000 8.00830.0500 8.00830.0500
EM8.00830.5000EA 333 USD InStock
CA8.00830.5000 EM8.00830.5000 CA8.00830.0500 EM8.00830.0500 CA8.00830.0100 EM8.00830.0100
Acrylamide for synthesis, Sigma-Aldrich®
Acrylamide (monomer)


Formula: C₃H₅NO
MW: 71.08 g/mol
MDL Number: MFCD00008032
CAS Number: 79-06-1
UN: 2074
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg
Order Now
Specification Test Results
Solubility 2040 g/l (25°C)
Melting Point 84 °C
Molar Mass 500 kg/m3
Bulk Density 500 kg/m3
Boiling Point 125 °C (33.3 hPa)
Vapor Pressure 0.009 hPa (25 °C)
Density 1.127 g/cm3 (25 °C)
pH 5.0 - 7.0 (50 g/l, H2O, 20 °C)
Ignition Point 424°C
Synonyms Prop-2-enamide
Hill Formula C3H5NO
Chemical Formula CH2CHCONH2

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty